Learn More
Invitrogen™ SERPINC1 Polyclonal Antibody
Artikelnummer. 10322570
Change view
Click to view available options
Menge:
400 μL
Packungsgröße:
100 Mikrogramm
Artikelnummer. Menge unitSize
10322570 400 μL 100 Mikrogramm
1 options
Artikelnummer. 10322570
Marke: Invitrogen™ PA513674
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rabbit Polyclonal Antibody
SERPINC1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
The protein encoded by this gene is a plasma protease inhibitor and a member of the serpin superfamily. This protein inhibits thrombin as well as other activated serine proteases of the coagulation system, and it regulates the blood coagulation cascade. The protein includes two functional domains: the heparin binding-domain at the N-terminus of the mature protein, and the reactive site domain at the C-terminus. The inhibitory activity is enhanced by the presence of heparin. More than 120 mutations have been identified for this gene, many of which are known to cause antithrombin-III deficiency.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen SERPINC1
Anwendungen Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Klassifikation Polyclonal
Konzentration 1.8 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 0.09% sodium azide; pH 7.4
Gen Serpinc1
Gen-Zugriffsnummer P01008, P32261
Gen-Alias AI114908; Antithrombin; Antithrombin 3; anti-thrombin 3; antithrombin III; antithrombin-III; AT3; At-3; AT3D; ATIII; hypothetical protein LOC540261; MGC22579; PRO0309; serine (or cysteine) peptidase inhibitor, clade C (antithrombin), member 1; serine (or cysteine) proteinase inhibitor, clade C (antithrombin), member 1; serpin C1; serpin family C member 1; serpin peptidase inhibitor clade C member 1; serpin peptidase inhibitor, clade C (antithrombin), member 1; serpin peptidase inhibitor, clade C, member 1; Serpinc1; THPH7
Gensymbole Serpinc1
Wirtsspezies Rabbit
Immunogen KLH conjugated synthetic peptide between 364-393 amino acids from the C-terminal region of human SERPINC1.
Reinigungsverfahren Ammonium sulfate precipitation, Size-exclusion - Dialysis
Menge 400 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 11905, 462
Zielspezies Human, Mouse
Inhalt und Lagerung Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotype IgG