Learn More
SERPIND1 Mouse anti-Human, CoraLite 488, Clone: 2C1E1, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL488-67602-100UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The SERPIND1 antibody from Proteintech is a mouse monoclonal antibody to a recombinant protein of human SERPIND1. This antibody recognizes human antigen. The SERPIND1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Spezifikation
| SERPIND1 | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 3053 | |
| Liquid |
| Immunofluorescence | |
| 2C1E1 | |
| Mouse | |
| Primary | |
| Human | |
| IgG1 |