missing translation for 'onlineSavingsMsg'
Learn More

SETD6 Antibody - BSA Free, Novus Biologicals™

Artikelnummer. 18604411 Alle ansehen Bio Techne Produkte
Ansicht ändern
Click to view available options
Menge:
0.02 mL
0.1 mL
Packungsgröße:
0.01 Milliliter
0.02 Milliliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
18604411 0.1 mL 0.01 Milliliter
18610161 0.02 mL 0.02 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 18604411 Lieferant Novus Biologicals Lieferanten-Nr. NBP2930310.1ml

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

SETD6 Polyclonal antibody specifically detects SETD6 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen SETD6
Anwendungen Western Blot
Klassifikation Polyclonal
Konjugat Unconjugated
Verdünnung Western Blot 1:500 - 1:1000
Zusammensetzung PBS (pH 7.3), 50% glycerol
Gen-Alias SET domain containing 6, SET domain-containing protein 6
Wirtsspezies Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 301-473 of human SETD6 (NP_001153777.1). EPYPDNTDDTADIQMVTVREAALQGTKTEAERHLVYERWDFLCKLEMVGEEGAFVIGREEVLTEEELTTTLKVLCMPAEEFRELKDQDGGGDDKREEGSLTITNIPKLKASWRQLLQNSVLLTLQTYATDLKTDQGLLSNKEVYAKLSWREQQALQVRYGQKMILHQLLELTS
Reinigungsverfahren Affinity purified
Menge 0.1 mL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 79918
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.