Learn More
Invitrogen™ Setrusumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Marke:  Invitrogen™ MA558770
 Weitere Artikel aus diesem Sortiment
Artikelnummer. 30283445
  • 1262.00 € / 1 Milligramm
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Setrusumab is under investigation in clinical trial NCT01406548 (Safety and Efficacy of Multiple Dosing Regimens of BPS804 in Post Menopausal Women With Low Bone Mineral Density).
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
Setrusumab
Recombinant Monoclonal
Unconjugated
BPS-804; MOR-05813
Protein A
RUO
Human
Antibody
IgG2SA
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized