Beschreibung
The SF3B3 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human SF3B3. This antibody recognizes human, mouse, rat antigen. The SF3B3 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Spezifikation
Spezifikation
| Antigen | SF3B3 |
| Anwendungen | Immunofluorescence |
| Konzentration | 1000 μg/mL |
| Konjugat | CoraLite 594 |
| Gensymbole | KIAA0017, RSE1, SAP 130, SAP130, SF3b130, SF3B3, Splicing factor 3B subunit 3, STAF130 |
| Wirtsspezies | Rabbit |
| Menge | 20 μL |
| Primär oder sekundär | Primary |
| Gen-ID (Entrez) | 23450 |
| Zielspezies | Human, Mouse, Rat |
| Mehr anzeigen |