missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SF3B3 Recombinant Rabbit Monoclonal Antibody (K01_1Y30)
Description
Immunogen sequence: SMEPNKQKNV SEELDRTP Highest antigen sequence identity to the following orthologs: Mouse - 100%, Cynomolgus monkey - 100%.
This gene encodes subunit 3 of the splicing factor 3b protein complex. Splicing factor 3b, together with splicing factor 3a and a 12S RNA unit, forms the U2 small nuclear ribonucleoproteins complex transcription coactivator-HAT complex, and the TFTC -containing complex). These complexes may function in chromatin modification, transcription, splicing, and DNA repair.
Specifications
Specifications
| Antigen | SF3B3 |
| Anwendungen | Western Blot, Immunocytochemistry |
| Klassifikation | Recombinant Monoclonal |
| Klon | K01_1Y30 |
| Konzentration | 1 mg/mL |
| Konjugat | Unconjugated |
| Zusammensetzung | 50mM tris glycine with 150mM NaCl, 0.05% BSA, 40% glycerol and 0.01% sodium azide; pH 7.4 |
| Gen | SF3B3 |
| Gen-Zugriffsnummer | Q15393, Q921M3 |
| Gen-Alias | 1810061H24Rik; 5730409A01Rik; AA409318; D8Ertd633e; Kiaa0017; mKIAA0017; pre-mRNA splicing factor SF3b, 130 kDa subunit; pre-mRNA-splicing factor SF3b 130 kDa subunit; RSE1; SAP 130; SAP130; SF3b130; SF3B3; Spliceosome-associated protein 130; splicing factor 3b subunit 3; splicing factor 3b, subunit 3; STAF130 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?