Learn More
Micronova™ X12 Pen, Sharpie Pro Extra Fine Point Marker, Bl
Limits particle generation with special poly barrel. Micronova™l Sharpie Pro™ Extra Fine Point Cleanroom Marker dries quickly and is permanent on most surfaces.
Marke: Micronova™ PEN-50IR
Weitere Details : Netto-Gewicht : 0.20000kg
Beschreibung
- Sterile cleanroom markers with extra-fine tip
- Marks on most surfaces
- Permanent fade and water resistant black ink
- Quick drying, non-toxic alcohol based ink formula
- Individually bagged and gamma-irradiated
Spezifikation
Cleanroom Marker | |
Black | |
Marking on most surfaces | |
Individually bagged and irradiated |
Extra Fine | |
Sterile | |
12 Pk. |