Learn More
SHBG Antibody - BSA Free, Novus Biologicals™
Artikelnummer. 18716753 Alle ansehen Bio Techne Produkte
Change view
Click to view available options
Menge:
0.025 mg
0.1 mg
Packungsgröße:
0.03 Milligramm
0.10 Milligramm
Artikelnummer. Menge unitSize
18716753 0.1 mg 0.10 Milligramm
18637676 0.025 mg 0.03 Milligramm
2 options
Artikelnummer. 18716753
Marke: Novus Biologicals NBP241294
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Rabbit Polyclonal Antibody
SHBG Polyclonal specifically detects SHBG in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Spezifikation
Antigen SHBG
Anwendungen Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Klassifikation Polyclonal
Konzentration 1 mg/mL
Konjugat Unconjugated
Verdünnung Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 20 ug/mL
Zusammensetzung PBS with 0.02% Sodium Azide
Gen-Alias ABPTestosterone-estrogen-binding globulin, androgen binding protein, MGC126834, MGC138391, SBP, sex hormone-binding globulin, Sex steroid-binding protein, TeBG, Testis-specific androgen-binding protein, testosterone-binding beta-globulin, Testosterone-estradiol-binding globulin
Gensymbole SHBG
Wirtsspezies Rabbit
Immunogen Antibody was raised against a 16 amino acid peptide near the center of human SHBG. The immunogen is located within amino acids 220 - 270 of SHBG. Amino Acid Squence: AEFNLRDIPQPHAEPW
Molekulargewicht des Antigens 35 kDa
Reinigungsverfahren Affinity Purified
Menge 0.1 mg
Regulatorischer Status RUO
Forschungsgebiet Cancer, Vision
Primär oder sekundär Primary
Gen-ID (Entrez) 6462
Testspezifität At least three isoforms of SHBG are known to exist; this antibody will detect all three isoforms.
Zielspezies Human, Mouse, Rat
Inhalt und Lagerung Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG