missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SIGLEC6 Monoclonal Antibody (1004337)
Mouse Monoclonal Antibody
Marke: Invitrogen™ MA561212
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.5 mg/mL in sterile PBS.
SIGLEC9 (Sialic acid-binding Ig-like lectin 9) is a putative adhesion molecule that mediates sialic-acid dependent binding to cells. It contains two Ig-like C2 type (immunoglobulin-like) domains and one Ig-like V-type (immunoglobulin-like) domain. It preferentially binds to alpha2, 3-or alpha2, 6-linked sialic acid. The sialic acid recognition site may be mased by cis interactions with sialic acids on the same cell surface.
Spezifikation
| SIGLEC6 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| O43699 | |
| SIGLEC6 | |
| Chinese Hamster Ovary cell line, CHO-derived human Siglec-6, Gln27-Ser331. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin) | |
| 1004337 | |
| Unconjugated | |
| SIGLEC6 | |
| CD327; CD33 antigen-like 1; CD33L; CD33L1; CD33L2; CDW327; OBBP1; OB-BP1; obesity-binding protein 1; sialic acid binding Ig like lectin 6; sialic acid binding Ig-like lectin 6; sialic acid-binding Ig-like lectin 6; Siglec; SIGLEC6; Siglec-6 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 946 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur