Learn More
Buerkle™ Silicone Hose
Artikelnummer. 17997790 Alle ansehen Bürkle Produkte
Click to view available options
Innendurchmesser (metrisch):
3 mm
5 mm
6 mm
Außendurchmesser (metrisch):
5 mm
8 mm
9 mm
Packungsgröße:
5 Meter
Artikelnummer. 17997790
Marke: Buerkle™ 88030305
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
This silicone tubing is the ideal choice for laboratory or food applications, it is physiologically safe and guarantees the highest quality and safety standards. The tubing can be sterilised in an autoclave or with ethylene oxide.
This silicone tubing is the ideal choice for laboratory or food applications, it is physiologically safe and guarantees the highest quality and safety standards. The tubing can be sterilised in an autoclave or with ethylene oxide.
Thanks to its high flexibility, the tubing is particularly adaptable and remains permanently elastic even at high and low temperatures.
Our high-quality silicone tubing is resistant to weak acids and alkalis as well as many other chemicals, making it suitable for a wide range of applications.
The transparency of the hose enables optimum visual control of the process.
  • Permanently elastic at high and low temperatures
  • Good transparency for visual inspection
  • Ideal for applications in food and food technology
TRUSTED_SUSTAINABILITY
Spezifikation
Autoklavierbar Autoclavable
Innendurchmesser (angloamerikanisch) 0.12 in.
Innendurchmesser (metrisch) 3 mm
Material Silicone
Außendurchmesser (angloamerikanisch) 0.20 in.
Außendurchmesser (metrisch) 5 mm
Gesamtlänge (angloamerikanisch) 16.4 ft.
Gesamtlänge (metrisch) 5 m
Farbe Transparent
Produkttyp Tubing