missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
Sin Nombre Virus Nucleocapsid Polyclonal antibody specifically detects Sin Nombre Virus Nucleocapsid in Virus samples. It is validated for ELISA
Spezifikation
Spezifikation
| Antigen | Sin Nombre Virus Nucleocapsid |
| Konjugat | PerCP |
| Reinigungsverfahren | Peptide affinity purified |
| Isotype | IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?