Learn More
SingleStep Poly-HRP anti-Mouse IgG (H+L) Secondary Antibody
Artikelnummer. 15921448
Change view
Click to view available options
Menge:
1 mg
10 mL
100 μg
100 mL
5 mL
5 mg
50 mL
500 μg
Packungsgröße:
1 Milligramm
10 Milliliter
100 Mikrogramm
100 Milliliter
5 Milligramm
5 Milliliter
50 Milliliter
500 Mikrogramm
Artikelnummer. Menge unitSize
15921448 5 mg 5 Milligramm
15971438 100 μg 100 Mikrogramm
15901448 500 μg 500 Mikrogramm
15991438 1 mg 1 Milligramm
15931448 5 mL 5 Milliliter
15981438 10 mL 10 Milliliter
15911448 50 mL 50 Milliliter
15961438 100 mL 100 Milliliter
8 options
Artikelnummer. 15921448
Marke: EnquireBio Q2AB15mg
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Dieser Artikel ist im Moment nicht verfügbar oder wird nicht mehr hergestellt.
Schauen Sie bitte auf unsere Produktseite nach möglichen Alternativen
Alternative Produkte anzeigen
Goat Polyclonal Antibody
SingleStep™ Poly-HRP (Horse Radish Peroxidase) conjugated goat anti-mouse secondary antibodies provide signal amplification over traditional HRP conjugated reagents in immunohistochemistry (IHC) staining using primary antibodies from a mouse host. The signal amplification afforded by these HRP polymer conjugated antibodies is sufficient to enable application of a single secondary antibody unlike standard HRP conjugated reagents which utilize an unconjugated or biotinylated secondary antibody and then an additional HRP conjugated secondary antibody or HRP conjugted streptavidin to visualize staining. These additional staining steps increase background staining and variability between samples which is obviously undesirable in IHC. SingleStep™ goat anti-mouse poly-HRP secondary antibodies are sourced from the industry leading secondary antibody manufacturer and are then conjugated to a carefully controlled number of HRP molecules to facilitate excellent tissue penetration without sacrificing signal amplification. Our predilute products also incorporate our enhanced IHC staining buffer which contains optimized stabilizers and blocking reagents to ensure maximum sensitivity and high reproducibility in IHC in most sample types.
Spezifikation
Antigen IgG (H+L)
Anwendungen Immunohistochemistry
Klassifikation Polyclonal
Konjugat HRP
Zusammensetzung Pre-dilute antibody is in proprietary signal enhancement buffer composed of protein based blocking agent (non-BSA), non-ionic detergent, preservative (non-azide), and HRP-stabilizer at pH 7.4.
Wirtsspezies Goat
Immunogen Generation of this anti-Poly-HRP Anti-Mouse IgG (H+L) Secondary Antibody antibody was achieved by immunizing a Goat host with Pooled population of Mouse IgG
Reinigungsverfahren Antibody is initially purified by ammonium sulfate precipitation followed by cross-adsorption to ensure minimal cross-reactivity to human, bovine, and equine antibodies. Purification of SingleStep(TM) poly-HRP conjugated goat anti-mouse secondary is achieved via size exclusion liquid chromatography.
Menge 5 mg
Regulatorischer Status RUO
Primär oder sekundär Secondary
Zielspezies Mouse
Inhalt und Lagerung Store at 4°C. Do not freeze. Stable for >1 year if stored properly.