Beschreibung
Sirtuin 1/SIRT1 Monoclonal antibody specifically detects Sirtuin 1/SIRT1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Spezifikation
Spezifikation
| Antigen | Sirtuin 1/SIRT1 |
| Anwendungen | Western Blot, Immunohistochemistry, Immunofluorescence |
| Klassifikation | Monoclonal |
| Klon | SR1063 |
| Konjugat | Unconjugated |
| Verdünnung | Western Blot 1:1000-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/Immunofluorescence 1:50-1:200 |
| Zusammensetzung | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Gen-Alias | EC 3.5.1, S. cerevisiae, homolog) 1, sirtuin 1 |
| Wirtsspezies | Rabbit |
| Immunogen | A synthesized peptide derived from human phospho-Sirtuin 1/SIRT1 [Ser47] (Uniprot #: Q96EB6) |
| Mehr anzeigen |