missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC11A2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 20507-1-AP-150UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
This antibody detected various forms of SLC11A2 of 45-100 kDa.
This gene encodes a member of the solute carrier family 11 protein family. The product of this gene transports divalent metals and is involved in iron absorption. Mutations in this gene are associated with hypochromic microcytic anemia with iron overload. A related solute carrier family 11 protein gene is located on chromosome 2. Multiple transcript variants encoding different isoforms have been found for this gene.Spezifikation
| SLC11A2 | |
| Polyclonal | |
| Unconjugated | |
| SLC11A2 | |
| DCT1, Divalent cation transporter 1, Divalent metal transporter 1, DMT 1, DMT1, NRAMP 2, NRAMP2, SLC11A2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 18174, 25715, 4891 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O54902, P49281, P49282 | |
| Slc11a2 | |
| SLC11A2 Fusion Protein Ag14314 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur