Learn More
Invitrogen™ SLC22A1 Recombinant Rabbit Monoclonal Antibody (24GB2510)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557256
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SLC22A1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Polyspecific organic cation transporters in the liver, kidney, intestine, and other organs are critical for elimination of many endogenous small organic cations as well as a wide array of drugs and environmental toxins. This gene is one of three similar cation transporter genes located in a cluster on chromosome 6. The encoded protein contains twelve putative transmembrane domains and is a plasma integral membrane protein. Two transcript variants encoding two different isoforms have been found for this gene, but only the longer variant encodes a functional transporter.
Spezifikation
| SLC22A1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SLC22A1 | |
| 1-Oct; DKFZp313N1010; FLJ25043; FLJ37489; H oct1_cds; hOCT1; Lx1; mOCT1; OCT1; oct1_cds; Orct; Orct1; organic cation transporter 1; rOCT1; Slc22a1; solute carrier family 22 (organic cation transporter), member 1; solute carrier family 22 member 1; solute carrier family 22, member 1; SYK | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 6580 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2510 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O15245 | |
| SLC22A1 | |
| A synthesized peptide derived from human SLC22A1 (aa 100-150). | |
| 20 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |