missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC25A19 Monoclonal Antibody (663323)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA561688
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
SLC25A19 encodes a mitochondrial protein that is a member of the solute carrier family. Although this protein was initially thought to be the mitochondrial deoxynucleotide carrier involved in the uptake of deoxynucleotides into the matrix of the mitochondria, further studies have demonstrated that this protein instead functions as the mitochondrial thiamine pyrophosphate carrier, which transports thiamine pyrophosphates into mitochondria.
Spezifikation
| SLC25A19 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q9DAM5 | |
| SLC25A19 | |
| Mouse MUP-1 synthetic peptide. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgM |
| Immunocytochemistry | |
| 663323 | |
| Unconjugated | |
| SLC25A19 | |
| 2900089E13Rik; DNC; LOC504418 protein; MCPHA; mitochondrial deoxynucleotide carrier; mitochondrial thiamine pyrophosphate carrier; Mitochondrial uncoupling protein 1; MUP1; Slc25a19; solute carrier family 25 (mitochondrial deoxynucleotide carrier), member 19; solute carrier family 25 (mitochondrial thiamine pyrophosphate carrier), member 19; solute carrier family 25 member 19; solute carrier family 25, member 19; THMD3; THMD4; TPC | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 67283 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur