missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLC4A7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA575807
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
SLC4A7 regulates intracellular pH and may also play a role in bicarbonate salvage in secretory epithelia. Additionally, SLC4A7 may also have associated sodium channel activity.
Spezifikation
| SLC4A7 | |
| Polyclonal | |
| Unconjugated | |
| SLC4A7 | |
| bicarbonate transporter; BT; E430014N10Rik; Electroneutral Na/HCO(3) cotransporter; electroneutral sodium bicarbonate cotransporter 1; Na-bicarbonate cotransporter; Nbc; NBC2; NBC2B; NBC3; NBC-like protein; Nbcn1; NBCn1-G; SBC2; SLC4A6; Slc4a7; Sodium bicarbonate cotransporter 2; sodium bicarbonate cotransporter 2b; Sodium bicarbonate cotransporter 3; sodium bicarbonate cotransporter NBCn1-A; sodium bicarbonate cotransporter NBCn1-B; sodium bicarbonate cotransporter NBCn1-c; sodium bicarbonate cotransporter NBCn1-D; sodium bicarbonate cotransporter NBCn1-H; solute carrier family 4 member 7; solute carrier family 4, sodium bicarbonate cotransporter, member 7 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 117955, 218756, 9497 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8BTY2, Q9R1N3, Q9Y6M7 | |
| SLC4A7 | |
| Synthetic peptide corresponding to residues in Human SLC4A7. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur