Learn More
Ibidi μ-Slide 18 Well ibiTreat
Beschreibung
- A chambered coverslip with 18 wells for cell culture, immunofluorescence, and high-end microscopy
- bottom material: #1.5 Polymer Coverslip
- tissue culture treated
- sterilized
Spezifikation
Spezifikation
| Typ | Chambered Coverglass |
| Sterilität | Steril |