Learn More
Ibidi μ-Slide 4 Well Glass Bottom
Beschreibung
- A chambered coverslip with 4 wells for use in TIRF and super-resolution microscopy
- bottom material: #1.5H (170 μm ± 5 μm) D 263 M Schott glass coverslip
- sterilized
Spezifikation
Spezifikation
| Typ | Chambered Coverglass |
| Wells | 4 |
| Sterilität | Steril |