Learn More
Ibidi μ-Slide 4 Well ibiTreat
Beschreibung
- A chambered coverslip with 4 wells for cell culture, immunofluorescence, and high-end microscopy
- bottom material: #1.5 Polymer Coverslip, tissue culture treated
- sterilized
Spezifikation
Spezifikation
| Typ | Chambered Coverglass |
| Wells | 4 |
| Sterilität | Steril |