Learn More
Ibidi μ-Slide 8 Well Grid-500 ibiTreat
Artikelnummer. 17216213
Encompass_Preferred
Change view
Click to view available options
:
μ-Slide
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer.
17216213 μ-Slide
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 17216213 Lieferant Ibidi Lieferanten-Nr. 80826G500
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
μ-Slide 8 Well Grid-500 ibiTreat is a chambered coverslip with 8 wells and an imprinted 500 μm cell location grid.
  • A chambered coverslip with 8 wells and an imprinted 500 μm cell location grid for use in immunofluorescence and high-end microscopy
  • bottom material: #1.5 Polymer Coverslip
  • tissue culture treated
  • sterilized
Spezifikation
Typ Chambered Coverglass
Wells 8
Sterilität Steril