Learn More
Ibidi μ-Slide III 3D Perfusion ibiTreat
μ-Slide III 3D Perfusion ibiTreat is a three channel μ-Slide used for the cultivation and perfusion of 3D cell structure.
Marke: Ibidi 80376
Weitere Details : Netto-Gewicht : 0.17900kg
Beschreibung
- A three channel slide used for the cultivation and perfusion of cells or spheroids in 3D matrices
- easy to connect with tubing and pump systems via female Luers
- bottom material: #1.5 Polymer Coverslip
- tissue culture treated
- sterilized
Spezifikation
Microscope Slide | |
15 |