Learn More
Ibidi μ-Slide III 3D Perfusion ibiTreat
Beschreibung
- A three channel slide used for the cultivation and perfusion of cells or spheroids in 3D matrices
- easy to connect with tubing and pump systems via female Luers
- bottom material: #1.5 Polymer Coverslip
- tissue culture treated
- sterilized
Spezifikation
Spezifikation
| Typ | Microscope Slide |
| Menge | 15 |