Learn More
Ibidi μ-Slide Spheroid Perfusion Bioinert
Beschreibung
- A perfusable channel slide with 3 x 7 wells for long-term spheroid cultivation and high-end microscopy
- easy to connect with tubing and pump systems via female Luers
- bottom material: #1.5 Polymer Coverslip with passivated surface (biologically inert)
- sterilized
Spezifikation
Spezifikation
| Typ | Microscope Slide |
| Menge | 15 |