Learn More
Invitrogen™ SLIT2 Recombinant Rabbit Monoclonal Antibody (24GB2210)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557252
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SLIT2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
SLIT2 are ligands for Robo receptors and play a vital role for axonal migration/navigation during neural development. A number of cleavage products through alternate splicing are reported in the literature for SLIT2 proteins. The C-terminal cleavage proteins are more diffusible than the larger N-terminal protein that is more tightly cell associated. SLIT2 protein is expressed in tissues such as fetal lung, kidney, and adult spinal cord.
Spezifikation
| SLIT2 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SLIT2 | |
| downregulated during adipocyte differentiation-1; Drad-1; E030015M03Rik; E130320P19Rik; mKIAA4141; neurogenic extracellular slit protein; SLIL3; Slit; slit guidance ligand 2; slit homolog 2 (Drosophila); slit homolog 2 protein; Slit homolog 2 protein C-product; Slit homolog 2 protein N-product; SLIT2; Slit-2; Slit2 protein | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20563, 360272, 9353 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2210 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| O94813, Q9R1B9, Q9WVC1 | |
| SLIT2 | |
| A synthesized peptide derived from human Slit2 (aa 1400-1500). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |