missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SLITRK1 Monoclonal Antibody (327829)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA561242
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute at 0.5 mg/mL in sterile PBS.
SLIT and NTRK-like family 1 (Slitrk1) is a member a protein family consisting of six homologous transmembrane proteins (Slitrk1-6) that share two conserved leucine-rich repeat domains in the extracellular domain and have significant homology to Slit, a secreted axonal growth-controlling protein. These proteins are also homologous to trk neurotrophin receptors in their intracellular domains. Expression of Slitrk proteins is highly restricted to neural and brain tumor tissues, but varies within the family. For example, Slitrk1 is expressed primarily in mature neurons. Overexpression of Slitrk1 in transfected neuronal cells induced unipolar neurites, while expression of the other Slitrk proteins inhibited neurite outgrowth, suggesting that these proteins are involved in the control of neurite outgrowth. While Slitrk1 variants have been suggested associated with Tourette's Syndrome, it is thought to play only a minor role if at all.
Spezifikation
| SLITRK1 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| Q96PX8 | |
| SLITRK1 | |
| Mouse myeloma cell line NS0-derived recombinant human SLITRK1, Ala16-Ser616. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunohistochemistry (Paraffin) | |
| 327829 | |
| Unconjugated | |
| SLITRK1 | |
| 3200001I04Rik; KIAA1910; Leucine-rich repeat-containing protein 12; LRRC12; RP11-395N17.1; SLIT and NTRK like family member 1; SLIT and NTRK-like family, member 1; SLIT and NTRK-like protein 1; slit and trk like gene 1; SLITRK1; Sltk1; TTM; UNQ233/PRO266 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 114798 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur