missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smad2 Antibody (CL8472) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP2-88916CL1
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Smad2 Monoclonal antibody specifically detects Smad2 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Spezifikation
| Smad2 | |
| Monoclonal | |
| CoraFluor 1 | |
| hMAD-2, hSMAD2, JV18, JV18-1SMAD, mothers against DPP homolog 2 (Drosophila), MAD homolog 2, Mad protein homolog, Mad, mothers against decapentaplegic homolog 2, MADH2MGC34440, MADR2MGC22139, Mad-related protein 2, mother against DPP homolog 2, mothers against decapentaplegic homolog 2, Mothers against DPP homolog 2, Sma- and Mad-related protein 2, SMAD 2, SMAD family member 2MAD, mothers against decapentaplegic homolog 2 (Drosophila), SMAD, mothers against DPP homolog 2, Smad2 | |
| This antibody was developed using a synthetic peptide derived from Q15796, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Metastasis, Phospho Specific, Stem Cell Signaling Pathway, Wnt Signaling Pathway | |
| 4087 | |
| Store at 4⪚C in the dark. Do not freeze. | |
| IgG2a |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL8472 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur