missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Smad4 Antibody (CL9774) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Marke: Novus Biologicals NBP3-05507JF669
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Smad4 Monoclonal antibody specifically detects Smad4 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
Spezifikation
| Smad4 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| DPC4SMAD, mothers against DPP homolog 4 (Drosophila), hSMAD4, JIP, MAD homolog 4, MADH4mothers against decapentaplegic homolog 4, mothers against decapentaplegic homolog 4, mothers against decapentaplegic, Drosophila, homolog of, 4, SMAD 4, SMAD family member 4MAD, mothers against decapentaplegic homolog 4 (Drosophila), SMAD, mothers against DPP homolog 4, Smad4 | |
| This antibody was developed using a recombinant protein derived from Q13485, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Signal Transduction, Stem Cell Signaling Pathway | |
| 4089 | |
| Store at 4⪚C in the dark. | |
| IgG1 |
| Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin) | |
| CL9774 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur