missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMARCB1 Recombinant Rabbit Monoclonal Antibody (8A7O2)

Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557894
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: LWRRLATVEE RKKIVASSHG KKTKPNTKDH GYTTLATSVT LLKASEVEEI LDGNDEKYKA VSISTEPPTY LREQKAKRNS QWVPTLPNSS HHLDAVPCST T.
The SWI-SNF complex is involved in the activation of transcription via the remodeling of nucleosome structure in an ATP-dependent manner. Brm (also designated SNF2alpha) and Brg-1 (also designated SNF2beta) are the ATPase subunits of the mammalian SWI-SNF complex. Brm, Brg-1, Ini1 (integrase interactor 1, also designated SNF5), BAF155 (also designated SRG3) and BAF170 are thought to comprise the functional core of the SWI-SNF complex. Addition of Ini1, BAF155 and BAF170 to Brg-1 appears to increase remodeling activity. Other complex subunits are thought to play regulatory roles. hSNF2L and hSNF2H both appear to be homologs of Drosophila ISWI, a Brm related ATPase that is present in chromatin remodeling complexes other than SWI/SNF, including the NURF (nucleosome remodeling factor).
Spezifikation
| SMARCB1 | |
| Recombinant Monoclonal | |
| 1.86 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q12824, Q9Z0H3 | |
| SMARCB1 | |
| A synthetic peptide corresponding to a sequence within amino acids 50-150 of human SMARCB1/SNF5 (NP_003064.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| ELISA, Western Blot | |
| 8A7O2 | |
| Unconjugated | |
| SMARCB1 | |
| AU020204; Baf47; BRG1-associated factor 47; BRG1-associated factor 47 (BAF47); CSS3; hSNF5; hSNFS; INI1; Integrase interactor 1 protein; malignant rhabdoid tumor suppressor; MRD15; mSNF5; PPP1R144; protein phosphatase 1, regulatory subunit 144; RDT; RTPS1; Sfh1p; SMARCB1; smarcb1a; SNF5; SNF5 homolog; SNF5/INI1; SNF5L1; Snr1; Sucrose nonfermenting yeast homolog like 1; sucrose nonfermenting, yeast, homolog-like 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1 isoform a; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1a; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily B member 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily B member 1-A; SWI/SNF-related matrix-associated protein; SWI10; SWNTS1; Transcription factor TYE4; Transcription regulatory protein SNF5; TYE4; Unknown (protein for MGC:138029); zgc:92517 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 20587, 361825, 6598 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur