missing translation for 'onlineSavingsMsg'
Learn More

SMCHD1 Antibody (CL4265) [Alexa Fluor™ 647], R&D Systems™

Artikelnummer. 30297359 Alle ansehen Bio Techne Produkte
Ansicht ändern
Click to view available options
Menge:
0.1 mL
Packungsgröße:
0.10 Milliliter
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
30297359 0.1 mL 0.10 Milliliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30297359 Lieferant Novus Biologicals Lieferanten-Nr. NBP259045AF647
 Eingeschränkter Artikel
Verfügbar ab: 10-12-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

A Novus product, part of R&D Systems. Mouse Monoclonal Antibody

SMCHD1 Monoclonal antibody specifically detects SMCHD1 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen SMCHD1
Anwendungen Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence
Klassifikation Monoclonal
Klon CL4265
Konjugat Alexa Fluor 647
Zusammensetzung 50mM Sodium Borate
Gen-Alias DKFZp686O0631, KIAA0650, structural maintenance of chromosomes flexible hinge domain containing 1, structural maintenance of chromosomes flexible hinge domain-containing protein 1
Wirtsspezies Mouse
Immunogen This antibody was developed using a recombinant protein derived from A6NHR9, with the exact immunogen sequence remaining proprietary.
Reinigungsverfahren Protein A purified
Menge 0.1 mL
Regulatorischer Status RUO
Forschungsgebiet Protein Kinase
Primär oder sekundär Primary
Gen-ID (Entrez) 23347
Zielspezies Human
Inhalt und Lagerung Store at 4⪚C in the dark.
Produkttyp Antibody
Form Purified
Isotype IgG1
Produktlinie Novus
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.