missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SMPD3 Monoclonal Antibody (758612)

Mouse Monoclonal Antibody
Marke: Invitrogen™ MA561778
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Reconstitute with sterile PBS to a final concentration of 0.5 mg/mL.
Catalyzes the hydrolysis of sphingomyelin to form ceramide and phosphocholine. Ceramide mediates numerous cellular functions, such as apoptosis and growth arrest, and is capable of regulating these 2 cellular events independently. Also hydrolyzes sphingosylphosphocholine. Regulates the cell cycle by acting as a growth suppressor in confluent cells. Probably acts as a regulator of postnatal development and participates in bone and dentin mineralization.
Spezifikation
| SMPD3 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 5% trehalose and no preservative | |
| Q9NY59 | |
| SMPD3 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human SMPD3, Val2-Ala655. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 758612 | |
| Unconjugated | |
| SMPD3 | |
| 4631433G07Rik; AI427456; AW537966; Cca1; confluent 3Y1 cell-associated 1; confluent 3Y1 cell-associated protein 1; fro; neutral sphingomyelin phosphodiesterase 3; neutral sphingomyelinase 2; Neutral sphingomyelinase II; NSMASE2; nSMase-2; SMPD3; Sphingomyelin phosphodiesterase 3; sphingomyelin phosphodiesterase 3, neutral; sphingomyelin phosphodiesterase 3, neutral membrane; sphingomyelin phosphodiesterase 3, neutral membrane (neutral sphingomyelinase II) | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 55512 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur