Learn More
Tocris Bioscience™ SNIPER(ER)-87
Click to view available options
Menge:
5 mg
Packungsgröße:
5 Milligramm
Beschreibung
SNIPER(ER)-87 is a potent and selective estrogen receptor α (ERα) Degrader (Specific and Nongenetic IAP-dependent Protein Erasers or SNIPER) (DC50 = 3 nM). SNIPER(ER)-87 comprises the IAP binding ligand LCL 161 (Cat .No. 7178) joined by a linker to the estrogen-binding moiety 4-hydroxytamoxifen (Cat. No. 3412). The product decreases ERα levels in breast cancer cells in vitro and in mouse ovaries in vivo. SNIPER(ER)-87 also reduces ERα levels and inhibits tumor proliferation in breast cancer xenograft mouse models.
Spezifikation
Spezifikation
| Chemischer Name oder Material | (2 S)-N-[(1 S)-1-Cyclohexyl-2-[(2 S)-2-[4-[3-[[14-[4-[1-(4-hydroxyphenyl)-2-phenyl-1-buten-1-yl]phenoxy]-12-methyl-11-oxo-3,6,9-trioxa-12-azatetradec-1-yl]oxy]benzoyl]-2-thiazolyl]-1-pyrrolidinyl]-2-oxoethyl]-2-(methylamino)propanamide |
| CAS | 2222354-91-6 |
| Menge | 5 mg |
| Ziel | Active Degraders |
| Summenformel | C59H73N5O10S |
| Reinheit | 0.98 |