Learn More
Invitrogen™ SNRK Recombinant Rabbit Monoclonal Antibody (HL2225)
GREENER_CHOICE
Rabbit Recombinant Monoclonal Antibody
Marke:  Invitrogen™ MA556878
Artikelnummer. 30276286
  • 514.00 € / 100 Mikroliter
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
Beschreibung
Beschreibung
SNRK Recombinant Monoclonal Antibody for Western Blot
SNRK may play a role in hematopoietic cell proliferation or differentiation. SNRK is the potential mediator of neuronal apoptosis.
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
SNRK
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
Q63553, Q9NRH2
SNRK
Recombinant fragment of human SNRK.
100 μL
Primary
Human, Rat
Antibody
IgG
Western Blot
HL2225
Unconjugated
SNRK
2010012F07Rik; AI448042; AW547029; E030034B15; HSNFRK; KIAA0096; mKIAA0096; R74830; RGD:69653}; SNF related kinase; SNF-1 related kinase; SNF1-related kinase; SNF-related serine/threonine-protein kinase; SNF-related serine/threonine-protein kinase; LOW QUALITY PROTEIN: SNF-related serine/threonine-protein kinase; SNFRK; SNRK; snrk {ECO:0000312
Rabbit
Protein A
RUO
170837, 54861
-20°C, Avoid Freeze/Thaw Cycles