Learn More
Invitrogen™ SNX1 Recombinant Rabbit Monoclonal Antibody (24GB2595)
Rabbit Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA557733
Beschreibung
SNX1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This gene encodes a member of the sorting nexin family. Members of this family contain a phox (PX) domain, which is a phosphoinositide binding domain, and are involved in intracellular trafficking. This endosomal protein regulates the cell-surface expression of epidermal growth factor receptor. This protein also has a role in sorting protease-activated receptor-1 from early endosomes to lysosomes. This protein may form oligomeric complexes with family members. This gene results in three transcript variants encoding distinct isoforms.
Spezifikation
| SNX1 | |
| Recombinant Monoclonal | |
| Unconjugated | |
| SNX1 | |
| HsT17379; MGC8664; Snx1; SNX1A; sorting nexin 1; sorting nexin 1A; sorting nexin-1; VPS5 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 6642, 84471 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB2595 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q13596, Q99N27 | |
| SNX1 | |
| A synthesized peptide derived from human SNX1 (aa 50-100). | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |