missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Natriummolybdat-Dihydrat, TRC
Spezifikation
Spezifikation
| Chemischer Name oder Material | Sodium Molybdate Dihydrate |
| CAS | 10102-40-6 |
| Menge | 100 g |
| Summenformel | Mo O4 . 2 Na . 2 H2 O |
| Synonym | Sodium molybdate dihydrate, Molybdate (MoO42-)','sodium','hydrate (1:2:2)','(T-4)- (9CI), Molybdic acid (H2MoO4)','disodium salt','dihydrate (8CI), Disodium molybdate (Na2MoO4) dihydrate, Disodium molybdate dihydrate, Molyhibit 100, Sodium molybdate (Na2MoO4) dihydrate, Sodium molybdate (Na2MoO4.2H2O) |
| InChI-Formel | InChI=1S/Mo.2Na.2H2O.4O/h;;;2*1H2;;;;/q;2*+1;;;;;2*-1 |
| SMILES | O.O.[Na+].[Na+].[O-][Mo](=O)(=O)[O-] |
| IUPAC-Name | Dinatrium; Dioxido(dioxo)molybdän; Dihydrat |
| Molekulargewicht (g/mol) | 241.95 |
| Formelmasse | 243.8857 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?