Learn More
Melford Sodium Phosphate Dibasic, Anhydrous
Beschreibung
Sodium Phosphate Dibasic Anhydrous is a chemical compound that is commonly known as disodium hydrogen phosphate or disodium phosphate.
This compound is an anhydrous form of sodium phosphate, meaning it does not contain any water molecules.
In the life science field, Sodium Phosphate Dibasic Anhydrous is primarily used as a buffering agent, which helps maintain a stable pH in various biological and biochemical processes.
It is often utilized in laboratory settings for the preparation of various biological reagents, cell culture media, and protein purification processes. The compound's buffering properties make it valuable in the formulation of diagnostic kits and pharmaceutical products, where maintaining a specific pH is critical for the stability and effectiveness of the final product
Spezifikation
Spezifikation
| Chemischer Name oder Material | Sodium Phosphate Dibasic, Anhydrous |
| CAS | 7558-79-4 |
| Menge | 1 kg |
| Summenformel | Na2HPO4 |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.