Learn More
Natriumtetraborat, wasserfrei, Puratronic™, 99.998 % (Metallbasis), Thermo Scientific Chemicals
Beschreibung
Sodium tetraborate is used as a co-catalyst for (2,2,6,6-tetramethylpiperidin-1-yl)oxyl and sodium hypochlorite oxidation of alcohols in greener and non-chlorinated solvents. It acts as a reagent for the decomposition of oxides and silicates by fusion. It is used to prepare buffers, which is used in gel electrophoresis of DNA. It acts as a source of borate, micronutrient fertilizer, preservative in taxidermy, additive in ceramic slips and glazes. It is also used as a precursor for sodium perborate monohydrate and boric acid. It finds application as fluoride detoxification, neutron absorber in nuclear reactors and to make indelible ink for dip pens.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Siedepunkt | 1575°C (decomposition) |
| Chemischer Name oder Material | Sodium tetraborate |
| Schmelzpunkt | 741°C |
| Namenshinweis | Anhydrous |
| Farbe | Weiß |
| Physikalische Form | Pulver |
| Menge | 25 g |
| Hinweise zum Prozentgehalt | (Metallbasis) |
| Strukturformel | Na2B4O7 |
| Merck Index | 14,8590 |
| Mehr anzeigen |
Sicherheit und Handhabung
- Gefahr
- Reproduktionstoxizität Kategorie 1B
- H360FD-Kann die Fruchtbarkeit beeinträchtigen. Kann das Kind im Mutterleib schädigen.
- P201-Vor Gebrauch besondere Anweisungen einholen.
- P280-Schutzhandschuhe/Schutzkleidung/Augenschutz/Gesichtsschutz tragen.
- P308+P313-BEI Exposition oder falls betroffen: Ärztlichen Rat einholen/ärztliche Hilfe hinzuziehen.
RUO – Research Use Only
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.