missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Sonepcizumab Recombinant Monoclonal Antibody

Recombinant Monoclonal Antibody
Marke: Invitrogen™ MA559795
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Sonepcizumab, also referred to as LT1009, is a humanized monoclonal antibody that targets sphingosine 1-phosphate (S1P), a signaling lipid active in promoting angiogenesis and tumor growth. Sphingosine-1-phosphate (S1P) is a critical lysolipid involved in multiple biological functions, including heart development, immune response, and hearing. It is a metabolite from cell membrane sphingolipids and is primarily regulated through its binding to specific G protein-coupled receptors named S1P receptors (S1PRs). These receptors are ubiquitously expressed in the human body, influencing various physiological and pathological processes. The signaling pathways initiated by S1P-S1PR interactions are crucial for immune cell trafficking, activation, and adaptive immune responses. Studies also highlight the role of S1P in regulating vascular stability, endothelial cell function, and as a mediator in inflammatory responses. The SPNS2 gene encodes a transporter protein essential for the active secretion of S1P, further emphasizing its relevance in maintaining physiological homeostasis.
Spezifikation
| Sonepcizumab Humanized | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur