Beschreibung
Sortilin Monoclonal antibody specifically detects Sortilin in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
Spezifikation
Spezifikation
| Antigen | Sortilin |
| Anwendungen | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence |
| Klassifikation | Monoclonal |
| Klon | CL6526 |
| Konjugat | Biotin |
| Zusammensetzung | PBS |
| Gen-Alias | 100 kDa NT receptor, Glycoprotein 95, Gp95LDLCQ6, Neurotensin receptor 3, NT3NTR3, sortilin, sortilin 1 |
| Wirtsspezies | Mouse |
| Immunogen | This antibody was developed using a recombinant protein derived from Q99523, with the exact immunogen sequence remaining proprietary. |
| Reinigungsverfahren | Protein A purified |
| Mehr anzeigen |