Learn More
Sotorasib, MedChemExpress
Artikelnummer. 16435606
Encompass_Preferred
Change view
Click to view available options
Menge:
5 mg
10 mg
50 mg
100 mg
Packungsgröße:
10 Milligramm
5 Milligramm
50 Milligramm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge unitSize
16435606 100 mg -
16491067 50 mg 50 Milligramm
16454808 10 mg 10 Milligramm
16427557 5 mg 5 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 16435606 Lieferant Medchem Express Lieferanten-Nr. HY114277
Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!
MedChemExpress Sotorasib (AMG-510) is a first-in-class, orally bioavailable, and selective KRAS G12C covalent inhibitor. Sotorasib irreversibly inhibits KRAS G12C by locking it in an inactive GDP-bound state. Sotorasib is the first KRAS G12C inhibitor in clinical development and leads to the regression of KRAS G12C tumors.
Sotorasib (AMG-510) is a first-in-class, orally bioavailable, and selective KRAS G12C covalent inhibitor. Sotorasib irreversibly inhibits KRAS G12C by locking it in an inactive GDP-bound state. Sotorasib is the first KRAS G12C inhibitor in clinical development and leads to the regression of KRAS G12C tumors.
Spezifikation
Chemischer Name oder Material Sotorasib
Physikalische Form Powder
CAS 2296729-00-3
Farbe White
Menge 100 mg
Hinweise zur Reinheitsqualität Research
Summenformel C30H30F2N6O3
Zur Verwendung mit (Anwendung) Cancer-Kinase/protease
Synonym AMG-510
Löslichkeitsinformationen DMSO : 50 mg/mL (89.19 mM; Need ultrasonic) ∣H2O : 33.33 mg/mL (59.46 mM; ultrasonic and adjust pH to 11 with NaOH)
SMILES O=C(C=C)N1C[C@H](C)N(C2=NC(N(C3=C(C)C=CN=C3C(C)C)C4=C2C=C(F)C(C5=C(O)C=CC=C5F)=N4)=O)CC1
Molekulargewicht (g/mol) 560.59
Formelmasse 560.59
Reinheit (%) 98.0%
Güte Research