missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beschreibung
The SOX1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SOX1. This antibody recognizes human, mouse antigen. The SOX1 antibody has been validated for the following applications: WB, ELISA analysis.
Spezifikation
Spezifikation
| Antigen | SOX1 |
| Anwendungen | Western Blot, ELISA |
| Klon | 1C10H2 |
| Konzentration | 1000 μg/mL |
| Konjugat | Unconjugated |
| Wirtsspezies | Mouse |
| Immunogen | Recombinant Protein |
| Reinigungsverfahren | Protein G purification |
| Menge | 150 μL |
| Primär oder sekundär | Primary |
| Mehr anzeigen |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?