Learn More
SOX10 Antibody (CL4455) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Marke:  Novus Biologicals NBP2-59050CL1
Artikelnummer. 30300840
  • 1220.00 € / 0.10 Milliliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
SOX10 Monoclonal antibody specifically detects SOX10 in Feline,Human,Mouse samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunohistochemistry (Frozen),Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Spezifikation
Spezifikation
SOX10
Monoclonal
CoraFluor 1
MGC15649, PCWH, SRY (sex determining region Y)-box 10, SRY-related HMG-box gene 10, transcription factor SOX-10, WS4C, WS4mouse, human homolog of
This antibody was developed using a recombinant protein derived from P56693, with the exact immunogen sequence remaining proprietary.
0.1 mL
Primary
Feline, Human, Mouse
Purified
Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunocytochemistry/Immunofluorescence
CL4455
PBS
Mouse
Protein A purified
RUO
6663
Store at 4⪚C in the dark. Do not freeze.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications