Beschreibung
SOX21 Monoclonal antibody specifically detects SOX21 in Human,Mouse samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Spezifikation
Spezifikation
| Antigen | SOX21 |
| Anwendungen | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
| Klassifikation | Monoclonal |
| Klon | CL4688 |
| Konjugat | PE |
| Zusammensetzung | PBS |
| Gen-Alias | SOX25SOX-A, SOXA, SRY (sex determining region Y)-box 21, SRY-box 21, transcription factor SOX-21 |
| Wirtsspezies | Mouse |
| Immunogen | This antibody was developed using a synthetic peptide derived from Q9Y651, with the exact immunogen sequence remaining proprietary. |
| Reinigungsverfahren | Protein A purified |
| Mehr anzeigen |