Learn More
Invitrogen™ SPAG4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA561732
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: PLENEPKEML TLSEYHERVR SQGQQLQQLQ AELDKLHKEV STVRAANSER VAKLVFQRLN EDFVRKPDYA LSSVGASIDL QKTSHDYAD Highest antigen sequence identity to the following orthologs: Mouse - 89%, Rat - 89%.
The mammalian sperm flagellum contains two cytoskeletal structures associated with the axoneme: the outer dense fibers surrounding the axoneme in the midpiece and principal piece and the fibrous sheath surrounding the outer dense fibers in the principal piece of the tail. Defects in these structures are associated with abnormal tail morphology, reduced sperm motility, and infertility. In the rat, the protein encoded by this gene associates with an outer dense fiber protein via a leucine zipper motif and localizes to the microtubules of the manchette and axoneme during sperm tail development.
Spezifikation
| SPAG4 | |
| Polyclonal | |
| Unconjugated | |
| SPAG4 | |
| 1700041K21Rik; acrosomal protein ACR55; cancer/testis antigen 127; CT127; mKIAA4118; MNCb-0953; outer dense fiber-associated protein SPAG4; Sad1 and UNC84 domain containing 4; SPAG4; sperm antigen 4; sperm associated antigen 4; sperm tail protein; sperm-associated antigen 4 protein; SUN domain-containing protein 4; SUN4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6676 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9NPE6 | |
| SPAG4 | |
| Recombinant protein corresponding to Human SPAG4. Recombinant protein control fragment (Product #RP-100903). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |