missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals SPINeasy 96-Well DNA Pro Kit for Soil
Alle ansehen MP Biomedicals Produkte
Click to view available options
:
SPINeasy 96-Well DNA Pro Kit for Soil
Beschreibung
The SPINeasy 96-Well DNA Pro Kit for Soil has been developed to accommodate a wide range of soil types.
- Designed in 96-well spin plate format, allowing for high throughput and rapid sample processing.
- Enables efficient lysis of microbial population present in complex soil samples, including bacteria, fungi, viruses, and protists; ensuring comprehensive genomic DNA extraction.
- Compatible with various challenging soil types, including those with low biomass or high contamination levels.
- Streamlined workflow reduces processing time, enabling faster and convenient isolation of pure microbial genomic DNA in 90 minutes.
- Performance testing demonstrates its efficacy in extracting nucleic acids from soils with varying biomass levels, ranging from high to low.
- The kit provides optimal yield and purity for various types of soil samples. It displays an exceptional ability to extract gDNA from different types of soil.
Usage Statement: Research Use Only (RUO)
Key Applications:
- PCR
- qPCR amplification
Spezifikation
Spezifikation
| Format | Kit |
| Isolation Technology | Silica Membrane Technology |
| Anzahl Reaktionen | 96 Preps |
| Produkttyp | DNA Purification |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur