Learn More
Tuttnauer Spray Arm Monitor Chamber Sensor
Alle ansehen Tuttnauer Produkte
Click to view available options
:
Spray Arm Monitor; For use with: Tiva Washer
Beschreibung
- Special sensor to control the frequency of spray arm rotation inside the washing chamber.
- If the spray arm is blocked, the operator is alerted by an alarm.
- This ensures that all glassware is washed thoroughly.
Spezifikation
Spezifikation
| Material | Stainless Steel |
| Produkttyp | Sensor |
| Zur Verwendung mit (Geräte) | Tiva Washer |
| Menge | 1 Ea. |