missing translation for 'onlineSavingsMsg'
Learn More

SPTBN1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Artikelnummer. 16819204 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
150 μL
20 μL
Packungsgröße:
150 Mikroliter
20 Mikroliter
Artikelnummer. Menge unitSize
16819204 20 μL 20 Mikroliter
16809204 150 μL 150 Mikroliter
2 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Artikelnummer. 16819204

Marke: Proteintech 197221AP20UL

Bitte Sie sich an, um diesen Artikel zu kaufen. Sie benötigen ein Web-Konto? Registrieren Sie sich noch heute!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Rabbit Polyclonal Antibody

Spectrin (Sp), the most abundant of the erythrocyte membrane skeleton proteins, helps these cells maintain their characteristic biconcave shape while remaining flexible and elastic. Erythrocyte Sp is a heterodimer composed of a 280 kDa alpha subunit and a 246 kDa beta subunit which associate in a side-to-side, antiparallel configuration to form a 100 nm rod-like structure. Sp in other tissues may be composed of distinct but homologous alpha and beta subunits, sometimes referred to as fodrin. A newly introduced nomenclature designates the Sp subunits of the erythrocyte as alpha-1 and beta-1, and the fodrin subunits as alpha-2 and beta-2. Alternatively spliced forms of each are designated as epsilon-1, epsilon-2, etc. (e. g. beta-1 epsilon-1).

Spezifikation

Antigen SPTBN1
Anwendungen Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Klassifikation Polyclonal
Konzentration 0.17 mg/mL
Konjugat Unconjugated
Zusammensetzung PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gen SPTBN1
Gen-Zugriffsnummer Q01082, Q62261
Gen-Alias Beta II spectrin, beta-fodrin, betaSpII, ELF, Fodrin beta chain, Spectrin beta chain, brain 1, SPTB2, SPTBN1
Gensymbole SPTBN1
Wirtsspezies Rabbit
Immunogen Peptide
Reinigungsverfahren Antigen Affinity Chromatography
Menge 20 μL
Regulatorischer Status RUO
Primär oder sekundär Primary
Gen-ID (Entrez) 20742, 6711
Zielspezies Human, Mouse
Inhalt und Lagerung -20°C
Produkttyp Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.