Learn More
Invitrogen™ SQSTM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA595088
Beschreibung
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human COLO-320 whole cell, human A549 whole cell. IHC: rat brain tissue, human gastric cancer tissue, human lung cancer tissue, human oesophagus squama cancer tissue, human rectal cancer tissue, human sarcoma tissue, human cholangiocarcinoma tissue, human liver cancer tissue, mouse brain tissue, rat liver tissue. ICC/IF: A549 cell. Flow: A549 cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
This gene encodes a multifunctional protein that binds ubiquitin and regulates activation of the nuclear factor kappa-B signaling pathway. The protein functions as a scaffolding/adaptor protein in concert with TNF receptor-associated factor 6 to mediate activation of NF-kB in response to upstream signals. Alternatively spliced transcript variants encoding either the same or different isoforms have been identified for this gene. Mutations in this gene result in sporadic and familial Paget disease of bone.
Spezifikation
| SQSTM1 | |
| Polyclonal | |
| Unconjugated | |
| Sqstm1 | |
| A170; EBI3-associated protein of 60 kDa; EBI3-associated protein p60; EBIAP; FTDALS3; hypothetical protein 1; ORCA; OSF-6; Osi; OSIL; oxidative stress induced; oxidative stress induced like; p60; p62; p62B; PDB3; phosphotyrosine independent ligand for the Lck SH2 domain p62; phosphotyrosine-independent ligand for the Lck SH2 domain of 62 kDa; PKC-zeta-interacting protein; protein kinase C-zeta-interacting protein; sb:cb621; Sequestosome 1; sequestosome-1; Sqstm1; STAP; STONE14; ubiquitin-binding protein p62; Unknown (protein for MGC:127197); zgc:85784; ZIP; ZIP3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 113894, 18412, 8878 | |
| -20°C | |
| Lyophilized |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 500 μg/mL | |
| PBS with 5mg BSA and 0.05mg sodium azide, 0.05mg thimerosal | |
| O08623, Q13501, Q64337 | |
| Sqstm1 | |
| A synthetic peptide corresponding to a sequence at the N-terminus of human SQSTM1 (91-110aa KDDIFRIYIKEKKECRRDHR). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.