Learn More
Novus Biologicals™ SR-BI Peptide
Alle ansehen Bio Techne Produkte
Click to view available options
Menge:
0,1 mg
Packungsgröße:
0.10 Milligramm
Spezifikation
Spezifikation
| Gene ID (Entrez) | 949 |
| Konzentration | 1 mg/mL |
| Inhalt und Lagerung | Store at -80°C. Avoid freeze/thaw cycles. |
| Zur Verwendung mit (Anwendung) | This Peptide is Useful as a Blocking Peptide for NB400-104 |
| Gen-Alias | CD36 and LIMPII analogous 1, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1, CD36L1scavenger receptor class B type 1, CLA1 CD36 antigen-like 1, CLA-1 HDLQTL6, Collagen type I receptor, thrombospondin receptor-like 1, MGC138242, SCARB1, scavenger receptor class B type III, scavenger receptor class B, member 1, SR-B1, SRB1 scavenger receptor class B member 1, SRBI |
| Gensymbol | SCARB1 |
| Menge | 0,1 mg |
| Forschungskategorie | Breast Cancer, Cancer, Cardiovascular Biology, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction, Virology Bacteria and Parasites |
| Lagerungspuffer | Peptide dissolved in dH2O. Contains no BSA. |