Learn More
SRF Mouse anti-Human, CoraLite 594, Clone: 1G8F4, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech CL594-66742-100UL
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
The SRF antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human SRF. This antibody recognizes human antigen. The SRF antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Spezifikation
| SRF | |
| Monoclonal | |
| CoraLite 594 | |
| 100 μL | |
| 6722 | |
| IgG2a |
| Immunofluorescence | |
| 1G8F4 | |
| Mouse | |
| Primary | |
| Human |